Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-085

Supelco

AB-PINACA 5-Pentanoic acid metabolite

100 μg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$283.00

$283.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$283.00

About This Item

Empirical Formula (Hill Notation):
C18H24N4O4
CAS Number:
Molecular Weight:
360.41
UNSPSC Code:
41116107
NACRES:
NA.24

$283.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

AB-PINACA 5-Pentanoic acid metabolite is a major urinary metabolite of the synthetic cannabinoid AB-PINACA. Over the past couple of years, clusters of cases of severe toxic effects and deaths related to AB-PINACA use have increased in the United States. This certified reference solution is suitable for use in synthetic cannabinoid testing methods by LC/MS or GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service