Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Q-005

Supelco

Quetiapine carboxylic acid-D8 solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$354.00

$354.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$354.00

About This Item

Empirical Formula (Hill Notation):
C21H15D8N3O3S
Molecular Weight:
405.54
UNSPSC Code:
41116107
NACRES:
NA.24

$354.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

Quetiapine carboxylic acid is the significant urinary metabolite of quetiapine, marketed as the atypical antipsychotic Seroquel® for the treatment of depression, bipolar disorder, and schizophrenia. This new internal standard of quetiapine carboxylic acid is applicable to numerous LC/MS and GC/MS applications including quantitation of patient levels, forensic and forensic toxicology analysis, isotope dilution methods, and calibrator preparation.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Seroquel is a registered trademark of Astrazeneca UK
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service