Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-185

Supelco

Pyridine-2-aldehyde-DNPH solution

287 μg/mL in acetonitrile: DMSO (7:3), certified reference material, ampule of 2 mL, Cerilliant®

Synonym(s):

Pyridine-2-aldehyde-DNPH solution

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$313.00

$313.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
$313.00

About This Item

Empirical Formula (Hill Notation):
C12H9N5O4
CAS Number:
Molecular Weight:
287.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$313.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 2 mL

manufacturer/tradename

Cerilliant®

concentration

287 μg/mL in acetonitrile: DMSO (7:3)

format

single component solution

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service