Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-183

Supelco

Phosphatidylethanol 16:0/20:4 (PEth 16:0/20:4) solution

new

1 mg/mL in methanol (as free phosphate), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72O8P · NH4
Molecular Weight:
742.02

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol (as free phosphate)

format

single component solution

storage temp.

-65 to -96°C

General description

PEth 16:0/20:4, also known as phosphatidylethanol 16:0/20:4, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/20:4, in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/20:4, levels is performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service