Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-127

Supelco

Phendimetrazine tartrate tartrate salt solution

1.0 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$130.00

$130.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$130.00

About This Item

Empirical Formula (Hill Notation):
C12H17NO · C4H6O6
Molecular Weight:
341.36
UNSPSC Code:
41116107
NACRES:
NA.24

$130.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

An analytical reference standard suitable for use in LC/MS or GC/MS methods for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications. Phendimetrazine, a prodrug of phenmetrazine, is a stimulant drug of the morpholine chemical class used as an appetite suppressant marketed under the names Bontril, Adipost, Anorex-SR, Appecon, Melfiat, Obezine, Phendiet, Plegine, Prelu-2, or Statobex.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service