Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-089

Supelco

Pentedrone Norephedrine Metabolite hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO · HCl
CAS Number:
Molecular Weight:
215.72
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

Quality Level

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

NC(C(O)C1=CC=CC=C1)CCC.Cl

InChI

1S/C11H17NO.ClH/c1-2-6-10(12)11(13)9-7-4-3-5-8-9;/h3-5,7-8,10-11,13H,2,6,12H2,1H3;1H

InChI key

IIOOMPVSYKQIRA-UHFFFAOYSA-N

General description

This certified solution standard is suitable for use in synthetic cathinone testing methods by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. This synthetic cathinone biomarker is a primary urinary metabolite of Pentedrone. Pentedrone is a synthetic cathinone and stimulant widely found in numerous bath salt blends and party pills.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service