Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-104

Supelco

6β-Naltrexol-3-β-D-glucuronide

1.0 mg/mL (20% Water in Acetonitrile), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$414.00

$414.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$414.00

About This Item

Empirical Formula (Hill Notation):
C26H33NO10
CAS Number:
Molecular Weight:
519.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$414.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL (20% Water in Acetonitrile)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

SMILES string

O[C@@H]1[C@@H](C(O)=O)O[C@@H](OC2=C(O3)C([C@]([C@@H]3[C@H](O)CC4)(CCN5CC6CC6)[C@]4(O)[C@H]5C7)=C7C=C2)[C@H](O)[C@H]1O

InChI

1S/C26H33NO10/c28-13-5-6-26(34)15-9-12-3-4-14(35-24-19(31)17(29)18(30)21(37-24)23(32)33)20-16(12)25(26,22(13)36-20)7-8-27(15)10-11-1-2-11/h3-4,11,13,15,17-19,21-22,24,28-31,34H,1-2,5-10H2,(H,32,33)/t13-,15-,17+,18+,19-,21+,22+,24-,25+,26-/m1/s1

InChI key

MZOUDXNTQKTCTC-TXYMGXTCSA-N

General description

6ß-Naltrexol-3-ß-D-glucuronide is a primary urinary metabolite of naltrexone, an opiate drug used to counter the effects of opioid overdose. Naltrexone is sold under the trade names such as Revia, Depade and Vivitrol. This Certified Spiking Solution® is suitable for a variety of LC/MS or GC/MS applications, including forensic toxicology analysis, urine drug testing, and hydrolysis controls.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service