Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-096

Supelco

2,5-Dimethoxy-4-nitrophenethylamine-13C,D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

2C-N-13C,D3 hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C913CH11D3N2O4 · HCl
Molecular Weight:
266.70
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

General description

A psychedelic drug and phenethylamine of the 2C series, 2C-N or 2,5-Dimethoxy-4-nitrophenethylamine, produces intense hallucinations similar to those of LSD. This stable-labeled internal standard is suitable for quantitation of 2C-N levels in urine or other biological matrices by LC/MS or GC/MS for applications in forensic analysis, clinical toxicology, or urine drug testing. In Europe, 2C-N as well as other 2C amines, have reportedly been found as adulterants in some illicit drugs.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service