Skip to Content
MilliporeSigma
All Photos(1)

Documents

I-507

Supelco

Immunosuppressants CRM in Human Whole Blood (K2EDTA) solution

50-2000 ng/mL (Human Whole Blood (K2EDTA)), certified reference material, vial of 3 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.77

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Cyclosporin A, 2000 ng/mL
Everolimus, 50 ng/mL
Sirolimus, 50 ng/mL
Tacrolimus, 50 ng/mL

packaging

vial of 3 mL

manufacturer/tradename

Cerilliant®

concentration

50-2000 ng/mL (Human Whole Blood (K2EDTA))

format

multi-component solution

storage temp.

−70°C

General description

A Certified SpikingSolution® of routinely tested Immunosuppressants including Cyclosporin A,Everolimus, Sirolimus and Tacrolimus at concentrations of 50-2000 ng/mL inHuman Whole Blood. Immunosuppressants are drugs that inhibit or prevent theactivity of the immune system and are routinely tested in clinical LC-MS/MStesting applications. This multi-component matrix mix from Cerilliant® issuitable for critical quantitative applications in research and clinicalenvironments including: for use in preparation of controls, calibrators, andlinearity standards for LC-MS/MS assays; as system tuning/suitabilitystandards; linearity verification; and internal proficiency testing. Thestable, quantitative solution format in Human Whole Blood matrix provides significantbenefits to the user such as convenience of use, material savings, increasedefficiency of labor and reduced matrix effects. Certified concentration valuesfor each component provides increased efficiency from less time remaking poolsdue to dilution errors or rerunning samples.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service