Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-162

Supelco

1,24-Dihydroxyvitamin D3 solution

50 μg/mL (Ethanol with 0.1%BHT (degassed)), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

1αα,24R-Dihydroxyvitamin D3, Tacalcitol

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$331.00

$331.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$331.00

About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.24

$331.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

50 μg/mL (Ethanol with 0.1%BHT (degassed))

format

single component solution

storage temp.

-65 to -96°C

SMILES string

O[C@H]1C[C@@H](C(=C)\C(=C/C=C2/[C@H]3[C@@]([C@H](CC3)[C@@H](CC[C@@H](O)C(C)C)C)(CCC/2)C)\C1)O

InChI key

BJYLYJCXYAMOFT-RSFVBTMBSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service