Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-158

Supelco

HU-308 solution

1 mg/mL (Degassed Methanol), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$409.00

$409.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$409.00

About This Item

Empirical Formula (Hill Notation):
C27H42O3
CAS Number:
Molecular Weight:
414.62
UNSPSC Code:
41116107
NACRES:
NA.24

$409.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Methanol)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O(C)c1c(c(cc(c1)C(CCCCCC)(C)C)OC)[C@@H]2[C@H]3C([C@H](C3)C(=C2)CO)(C)C

InChI

1S/C27H42O3/c1-8-9-10-11-12-26(2,3)19-14-23(29-6)25(24(15-19)30-7)20-13-18(17-28)21-16-22(20)27(21,4)5/h13-15,20-22,28H,8-12,16-17H2,1-7H3/t20-,21+,22-/m0/s1

InChI key

CFMRIVODIXTERW-BDTNDASRSA-N

General description

HU-308 is a synthetic cannabinoid that acts as a potent, highly selective agonist for the CB2 receptor. HU-308 is evaluated to be effective against the cytokine release syndrome (CRS), that is seen in many diseases and conditions, including COVID-19. A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, forensic analysis and pharmaceutical research applications.
Snap-N-Spike®/Snap-N-Shoot®

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service