Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F-054

Supelco

o-Fluorofentanyl hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Synonym(s):

o-Fluorofentanyl Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27FN2O · HCl
Molecular Weight:
390.92
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

ortho-Fluorofentanyl, also known as 2-Fluorofentanyl, is an opioid analgesic being an analogue of fentanyl that is sold as designer drug on the illicit drug market since 2016. A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for ortho-Fluorofentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications.

Application

  • Analytical method development for forensic toxicology: Research demonstrates the application of o-Fluorofentanyl in the development of quantitative LC-MS/MS methods to analyze it alongside other synthetic opioids in postmortem blood. This method is crucial for forensic and pharmacological studies, enabling precise measurement of drug potency and efficacy in fatal overdose cases (Truver et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service