Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E-143

Supelco

(±)-Epinephrine hydrochloride solution

1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

(±)-Epinephrine hydrochloride, (±)-Adrenalin, 4-(1-Hydroxy-2-[methylamino]ethyl)-1,2-benzenediol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$91.80

$91.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$91.80

About This Item

Empirical Formula (Hill Notation):
C9H13NO3 · HCl
CAS Number:
Molecular Weight:
219.67
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$91.80


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol ((as free base))

format

single component solution

storage temp.

-10 to -25°C

SMILES string

OC(CNC)C1=CC=C(O)C(O)=C1.Cl

InChI

1S/C9H13NO3.ClH/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;/h2-4,9-13H,5H2,1H3;1H

InChI key

ATADHKWKHYVBTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Epinephrine, also known as adrenaline, is a marker for catecholamine-secreting cancers such as pheochromocytoma, paraganglioma, and neuroblastoma. This Certified Reference Material is suitable for LC-MS/MS monitoring of epinephrine levels in urine or plasma for diagnostic testing or endocrinology.
Snap-N-Spike®/Snap-N-Shoot®

Biochem/physiol Actions

Adrenergic receptor agonist.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service