Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-161

Supelco

C-Peptide solution

100 μg/mL (Protease-free 1x PBS), certified reference material, ampule of 0.1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C129H211N35O48
CAS Number:
Molecular Weight:
3020.26
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Protease-free 1x PBS)

technique(s)

liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

InChI

1S/C129H211N35O48/c1-58(2)42-77(155-113(195)74(31-39-102(184)185)151-127(209)105(65(15)16)162-114(196)71(25-32-88(131)167)145-95(174)54-141-126(208)104(64(13)14)161-115(197)72(26-33-89(132)168)150-118(200)80(45-61(7)8)156-121(203)83(48-103(186)187)159-112(194)73(30-38-101(182)183)148-107(189)67(18)143-109(191)69(130)24-36-99(178)179)111(193)138-50-93(172)135-49-92(171)136-55-98(177)163-40-20-22-86(163)124(206)140-51-94(173)142-66(17)106(188)137-52-96(175)146-84(56-165)122(204)157-81(46-62(9)10)119(201)152-75(27-34-90(133)169)128(210)164-41-21-23-87(164)125(207)160-78(43-59(3)4)116(198)144-68(19)108(190)154-79(44-60(5)6)117(199)149-70(29-37-100(180)181)110(192)139-53-97(176)147-85(57-166)123(205)158-82(47-63(11)12)120(202)153-76(129(211)212)28-35-91(134)170/h58-87,104-105,165-166H,20-57,130H2,1-19H3,(H2,131,167)(H2,132,168)(H2,133,169)(H2,134,170)(H,135,172)(H,136,171)(H,137,188)(H,138,193)(H,139,192)(H,140,206)(H,141,208)(H,142,173)(H,143,191)(H,144,198)(H,145,174)(H,146,175)(H,147,176)(H,148,189)(H,149,199)(H,150,200)(H,151,209)(H,152,201)(H,153,202)(H,154,190)(H,155,195)(H,156,203)(H,157,204)(H,158,205)(H,159,194)(H,160,207)(H,161,197)(H,162,196)(H,178,179)(H,180,181)(H,182,183)(H,184,185)(H,186,187)(H,211,212)/t66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,104-,105-/m0/s1

InChI key

VOUAQYXWVJDEQY-QENPJCQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

C-peptide, also known as the connecting peptide, is a 31-amino-acid polypeptide connecting insulin′s A-chain to its B-chain in the proinsulin molecule. Measurement of C-peptide blood serum levels can be used to distinguish between diabetes or hypoglycemia conditions with similar clinical features. This certified Snap-N-Spike® solution of the fully-intact c-peptide protein is suitable for use as starting material in calibrators and controls in LC-MS/MS c-peptide testing methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service