Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-138

Supelco

Codeine-6-ß-D-glucuronide-D3 solution

100 μg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26D3NO9
CAS Number:
Molecular Weight:
478.51
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol: water (2:8)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

O[C@H]([C@H]1O)[C@H](O)[C@@H](C(O)=O)O[C@H]1O[C@@H](C=C2)[C@H]3[C@](C2[C@H]4C5)(CCN4C([2H])([2H])[2H])C6=C5C=CC(OC)=C6O3

InChI

1S/C24H29NO9.Na/c1-25-8-7-24-11-4-6-14(32-23-18(28)16(26)17(27)20(34-23)22(29)30)21(24)33-19-13(31-2)5-3-10(15(19)24)9-12(11)25;/h3-6,11-12,14,16-18,20-21,23,26-28H,7-9H2,1-2H3,(H,29,30);/q;+1/p-1/t11-,12-,14-,16?,17-,18+,20-,21-,23+,24-;/m0./s1/i1D3;

InChI key

PXJIWCXZTGPMDS-WNCZEZBLSA-M

General description

An internal standard suitable for LC/MS or GC/MS applications with codeine-6ß-D-glucuronide such as forensic analysis, urine drug testing or clinical toxicology. Codeine-6ß-D-glucuronide is a major urinary metabolite of codeine, an opiate commonly used as an antitussive and analgesic for management of mild pain. This Certified Spiking Solution® provides added value in allowing for quantitation of codeine-6ß-D-glucuronide levels by LC-MS/MS without the need for hydrolysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service