Skip to Content
MilliporeSigma
All Photos(1)

Documents

C-131

Supelco

25C-NBOMe hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22ClNO3 · HCl
CAS Number:
Molecular Weight:
372.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC(OC)=C(CCNCC2=C(OC)C=CC=C2)C=C1OC.[H]Cl

InChI

1S/C18H22ClNO3.ClH/c1-21-16-7-5-4-6-14(16)12-20-9-8-13-10-18(23-3)15(19)11-17(13)22-2;/h4-7,10-11,20H,8-9,12H2,1-3H3;1H

InChI key

PMTKNKZAOWWYGW-UHFFFAOYSA-N

General description

A Certified Spiking Solution® suitable as starting material for calibrators or controls in GC/MS or LC/MS 25C-NBOMe testing methods for forensic analysis, clinical toxicology, or urine drug testing applications. 25C-NBOMe is a second generation 2C amine derived from attaching an NBOMe (2-methoxybenzyl) group to the amine group of 2C-C. Sold under street names such as Boom, C-Boom, and N-Bomb this synthetic phenethylamine produces vivid visual hallucinations similar to those of LSD.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service