Skip to Content
MilliporeSigma
All Photos(1)

Documents

B-094

Supelco

Busulfan solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6S2
CAS Number:
Molecular Weight:
246.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

InChI

1S/C6H14O6S2/c1-13(7,8)11-5-3-4-6-12-14(2,9)10/h3-6H2,1-2H3

InChI key

COVZYZSDYWQREU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Maximum a posteriori Bayesian methods out-perform non-compartmental analysis for busulfan precision dosing - This study underscores the importance of advanced pharmacokinetic modeling for Busulfan, an antineoplastic agent, enhancing dosing accuracy and therapeutic outcomes in clinical settings (Hughes et al., 2024).


  • Beneficial effects of adipose-derived stromal vascular fraction on testicular injury caused by busulfan - Investigates the protective effects of stem cell derivatives against Busulfan-induced gonadal toxicity, contributing to fertility preservation research in oncology patients (Hekimoglu et al., 2024).


  • Transcriptomic profile of human iPSC-derived podocyte-like cells exposed to a panel of xenobiotics - Discusses the cellular impact of Busulfan on kidney cell models, shedding light on its potential side effects and mechanisms of toxicity at a molecular level (Murphy et al., 2024).


  • Time-efficient germ cell transplantation from goldfish (Carassius auratus) into adult common carp (Cyprinus carpio) - This unique study uses Busulfan for germ cell depletion before transplantation, showcasing its utility in veterinary and aquatic biology research (Vigoya et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F

Flash Point(C)

6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service