Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B-049

Supelco

(R)-(-)-Bromo-DragonFLY hydrochloride solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12BrNO2 · HCl
CAS Number:
Molecular Weight:
330.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

BrC1=C2C(OC=C2)=C(C[C@@H](C)N)C3=C1OC=C3.Cl

InChI

1S/C13H12BrNO2.ClH/c1-7(15)6-10-8-2-4-17-13(8)11(14)9-3-5-16-12(9)10;/h2-5,7H,6,15H2,1H3;1H/t7-;/m1./s1

InChI key

YDIDKNSMQNPNFC-OGFXRTJISA-N

General description

Bromo-DragonFLY is an analog of the 2C amine family that also includes 2C-B, 2C-I, and 2C-E. This designer drug is an extremely potent hallucinogen with duration of action much longer than LSD. Toxicity of Bromo-DragonFLY appears to be fairly high for humans when taken in larger doses.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service