Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-145

Supelco

Acylcarnitines Mix 3 (C8:1,C10:1,C12:1,C14:1,C14:2,C16:1,C18:1,C18:2) solution

100-200 μg/mL (Methanol:1% aqueous formic acid (90:10) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$4,070.00

$4,070.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$4,070.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$4,070.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

trans-2-decenoyl-L-carnitine, 100 μg/mL
trans-2-dodecenoyl-L-carnitine, 100 μg/mL
trans-2-hexadecenoyl-L-carnitine, 100 μg/mL
cis,cis-9,12-octadecadienoyl-L-carnitine, 100 μg/mL
trans-2-octenoyl-L-carnitine, 100 μg/mL
oleoyl-L-carnitine, 200 μg/mL
cis,cis-5,8-tetradecandienoyl-L-carnitine, 100 μg/mL
trans-2-tetradecenoyl-L-carnitine, 100 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100-200 μg/mL (Methanol:1% aqueous formic acid (90:10) (v/v))

format

multi-component solution

storage temp.

−70°C

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In this article we provide a complete workflow of quantification and identification of acylcarnitines in dried blood spots (DBS) using stable isotope-labeled (SIL) acylcarnitines CRMs by FIA-MS/MS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service