Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-114

Supelco

AH-7921-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19D3Cl2N2O · HCl
Molecular Weight:
368.74
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(NCC1(N(C)C)CCCCC1)C2=C([2H])C(Cl)=C(Cl)C([2H])=C2[2H].Cl

General description

This stable-labeled internal standard is suitable for quantitation of AH-7921 levels in urine or blood by LC-MS or GC/MS methods in clinical toxicology, urine drug testing, or forensic analysis applications. AH-7921 is an opioid designer drug with 80% the potency of morphine when administered orally. First discovered in 2013, AH-7921 is an active ingredient of synthetic cannabis found in Japan.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Metzger et al.
Environmental science & technology, 50(15), 8188-8195 (2016-06-29)
The present study describes new procedures to obtain at millimeter resolution the spatial distribution of nitrite and nitrate in porewaters, combining diffusive equilibrium in thin films (DET), colorimetry and hyperspectral imagery. Nitrite distribution can be easily achieved by adapting the
Vladimir Zhemkov et al.
eLife, 10 (2021-05-12)
Sigma 1 receptor (S1R) is a 223-amino-acid-long transmembrane endoplasmic reticulum (ER) protein. S1R modulates activity of multiple effector proteins and is a well-established drug target. However, signaling functions of S1R in cells are poorly understood. Here, we test the hypothesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service