Skip to Content
MilliporeSigma
All Photos(1)

Documents

LM4100

Avanti

cholesterol (D7)

Avanti Research - A Croda Brand LM4100, methanol solution

Synonym(s):

cholest-5-en-3β-ol(d7)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39D7O
CAS Number:
Molecular Weight:
393.70
UNSPSC Code:
12352100
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM4100-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM4100

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3,18D

InChI key

HVYWMOMLDIMFJA-IFAPJKRJSA-N

Application

Cholesterol (D7) has been used in the preparation of liposome and as an internal standard to determine the retention time calibration.

Packaging

2 mL Amber Glass Sealed Ampule (LM4100-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct visualization of the conformational dynamics of single influenza hemagglutinin trimers
Das D, et al.
Cell, 174(4), 926-937 (2018)
Generation and quality control of lipidomics data for the alzheimer?s disease neuroimaging initiative cohort
Barupal DK, et al.
Scientific data, 5, 180263-180263 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service