Skip to Content
MilliporeSigma
All Photos(1)

Documents

900300M

Avanti

C32:6 fatty acid

Avanti Research - A Croda Brand 900300M, methylene chloride solution

Synonym(s):

14Z,17Z,20Z,23Z,26Z,29Z-dotriacontahexaenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H52O2
CAS Number:
Molecular Weight:
468.75
UNSPSC Code:
12352211
NACRES:
NA.25

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (900300M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900300M

concentration

1 mg/mL (900300M-1mg)

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C32H52O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31-32(33)34/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-31H2,1H3,(H,33,34)

InChI key

HIKGLCYKBLODNY-UHFFFAOYSA-N

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap with Foil Bag Argon Overpack (900300M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Poulos et al.
Advances in experimental medicine and biology, 318, 331-340 (1992-01-01)
Fatty acids with from 24 to 28 carbon atoms (very long-chain fatty acids, VLCFA) are present in small amounts in all mammalian tissues. Even longer chain fatty acids with from 30 to 38 carbon atoms (ultra-long-chain fatty acids, ULCFA) are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service