Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900220O

Avanti

Polyprenal (13~21)

Avanti Research - A Croda Brand 900220O

Synonym(s):

Polyprenal mixture (13~21)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mg (900220O-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900220O

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)=CCC/C(C)=C/CC/C(C)=C/CC/C(C)=C\CC/C(C)=C\C=O

Packaging

5 mL Clear Glass Sealed Ampule (900220O-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service