Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900119P

Avanti

PGF2β

Avanti Research - A Croda Brand

Synonym(s):

Prostaglandin F2β

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$145.00

$145.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$145.00

About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$145.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of (pkg of 1 X 1 mg (900119P-1MG))

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1C[C@@H](O)[C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/[C@@H](O)CCCCC

InChI key

PXGPLTODNUVGFL-JZFBHDEDSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (900119P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service