Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880700P

Avanti

16:0 PEG1000 PE

Avanti Research - A Croda Brand 880700P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-1000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$316.21

$316.21


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$316.21

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$316.21


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 200 mg (880700P-200mg)
pkg of 1 × 25 mg (880700P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880700P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C83H164NO32P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-81(85)113-78-80(116-82(86)31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)79-115-117(88,89)114-33-32-84-83(87)112-77-76-111-75-74-110-73-72-109-71-70-108-69-68-107-67-66-106-65-64-105-63-62-104-61-60-

InChI key

NLPYFLRBZNTJNT-MSPJXZLPSA-N

General description

Phosphoethanolamine serves as a substrate for several phospholipids of the cell membrane. It is associated with phospholipid turnover. Phosphoethanolamine is also found to prevent tumor progression.[1]

Application

16:0 PEG1000 PE is suitable for use as a component of nanoliposome.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (880700P-200mg)
5 mL Amber Glass Screw Cap Vial (880700P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoliposomes presenting on surface a cis-glycofused benzopyran compound display binding affinity and aggregation inhibition ability towards Amyloid $\beta$1-42 peptide
Airoldi C, et al.
European Journal of Medicinal Chemistry, 85, 43-50 (2014)
Adilson Kleber Ferreira et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 67(6), 481-487 (2013-06-19)
Phosphoethanolamine (Pho-s) is a compound involved in phospholipid turnover, acting as a substrate for many phospholipids of the cell membranes. In a recent study, we showed that Pho-s has antitumor effect in the several tumor cells. In this study we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service