880155P
Avanti
ALC-0159
Avanti Research™ - A Croda Brand
Synonym(s):
α-[2-(ditetradecylamino)-2-oxoethyl]-ω-methoxy-poly(oxy-1,2-ethanediyl), PEG-N,N-ditetradecylacetamide, Methoxypolyethyleneglycoloxy(2000)-N,N-ditetradecylacetamide
Select a Size
$433.61
Select a Size
About This Item
$433.61
Recommended Products
form
powder
packaging
pkg of 1x1 g (880155P-1G)
pkg of 1x100 mg (880155P-100MG)
pkg of 1x200 mg (880155P-200MG)
pkg of 1x25 mg (880155P-25MG)
pkg of 1x50 mg (880155P-50MG)
pkg of 1x500 mg (880155P-500MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
technique(s)
thin layer chromatography (TLC): suitable
application(s)
advanced drug delivery
lipid type
cationic lipids
polymerizable lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCN(CCCCCCCCCCCCCC)C(COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O
InChI
1S/C121H243NO47/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-122(29-27-25-23-21-19-17-15-13-11-9-7-5-2)121(123)120-169-119-118-168-117-116-167-115-114-166-113-112-165-111-110-164-109-108-163-107-106-162-105-104-161-103-102-160-101-100-159-99-98-158-97-96-157-95
InChI key
LPAYAXQFSOFRTA-UHFFFAOYSA-N
Packaging
- 5 mL Amber Glass Screw Cap Vial (880155P-50MG)
- 5 mL Amber Glass Screw Cap Vial (880155P-100MG)
- 5 mL Amber Glass Screw Cap Vial (880155P-200MG)
- 5 mL Amber Glass Screw Cap Vial (880155P-500MG)
- 30 mL Amber Glass Screw Cap Vial(880155P-1G)
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service