Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870818M

Avanti

N-arachidonoylglycine

Avanti Research - A Croda Brand 870818M, methylene chloride solution

Synonym(s):

methylene chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H35NO3
CAS Number:
Molecular Weight:
361.52
UNSPSC Code:
12352211
NACRES:
NA.25

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (870818M-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870818M

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)NCC(O)=O

InChI

1S/C22H35NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21(24)23-20-22(25)26/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-20H2,1H3,(H,23,24)(H,25,26)/b7-6-,10-9-,13-12-,16-15-

InChI key

YLEARPUNMCCKMP-DOFZRALJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-arachidonoylglycine (NAgly) is an endogenous ligand and a signaling lipid. It belongs to the eicosanoid family. NAgly is a natural ligand to activate G-protein coupled receptor, namely GPR18. It is found in rat brain and spinal cord.

Biochem/physiol Actions

N-arachidonoylglycine (NAgly) exhibits analgesic effects.

Packaging

2 mL Amber Glass Sealed Ampule (870818M-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Huang et al.
The Journal of biological chemistry, 276(46), 42639-42644 (2001-08-24)
In mammals, specific lipids and amino acids serve as crucial signaling molecules. In bacteria, conjugates of lipids and amino acids (referred to as lipoamino acids) have been identified and found to possess biological activity. Here, we report that mammals also
Resolution of inflammation by N-arachidonoylglycine
Burstein S H, et al.
Journal of Cellular Biochemistry, 112(11), 3227-3233 (2011)
T Sheskin et al.
Journal of medicinal chemistry, 40(5), 659-667 (1997-02-28)
In order to establish the structural requirements for binding to the brain cannabinoid receptor (CB1), we have synthesized numerous fatty acid amides, ethanolamides, and some related simple derivatives and have determined their Ki values. A few alpha-methyl- or alpha, alpha-dimethylarachidonoylalkylamides
S H Burstein et al.
Prostaglandins & other lipid mediators, 61(1-2), 29-41 (2000-04-29)
In addition to the well studied hydrolytic metabolism of anandamide, a number of oxidative processes are also possible. Several routes somewhat analogous to the metabolism of free arachidonic acid have been reported. These involve mediation by various lipoxygenases and COX-2
Identification of N-arachidonylglycine as the endogenous ligand for orphan G-protein-coupled receptor GPR18
Kohno M, et al.
Biochemical and Biophysical Research Communications, 347(3), 827-832 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service