Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870702P

Avanti

02:0 Coenzyme A

Avanti Research - A Croda Brand 870702P, powder

Synonym(s):

acetoyl Coenzyme A (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$591.00

$591.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$591.00

About This Item

Empirical Formula (Hill Notation):
C23H35N7Na3O17P3S
CAS Number:
Molecular Weight:
875.52
UNSPSC Code:
12352211
NACRES:
NA.25

$591.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870702P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870702P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(C)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[Na+].[Na+].[Na+]

InChI key

CRTIJMBXIFKWCA-JHJDYNLLSA-K

General description

This product is 90-95% pure 02:0 Coenzyme A with 5~10% free Coenzyme A.

Biochem/physiol Actions

Acetyl-CoA is an essential cofactor and carrier of acyl groups in enzymatic acetyl transfer reactions during protein post-translation. Oxidative decarboxylation of pyruvate yields acetyl CoA. It is also produced by condensation of acetate and thiol catalysed by acetyl CoA synthase. Oxidation of acetyl CoA in tricarboxylic acid cycle is associated with energy production.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870702P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epigenetic Cancer Therapy, 404-404 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service