Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870413C

Avanti

18:1 PE-benzylguanine

solution

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-benzylguanine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$361.00
5 MG
$1,080.00

$361.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$361.00
5 MG
$1,080.00

About This Item

Empirical Formula (Hill Notation):
C55H93N8O10P
CAS Number:
Molecular Weight:
1057.35
UNSPSC Code:
12352211
NACRES:
NA.25

$361.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99% (TLC)

form

solution

packaging

pkg of 1 × 1 mg (870413C-1MG)
pkg of 1 × 5 mg (870413C-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand (870413C)

concentration

1 mg/mL in chloroform
5 mg/mL in chloroform

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service