Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870287P

Avanti

16:0 Cyanur PE

Avanti Research - A Croda Brand 870287P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(cyanur) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H72Cl2N4NaO8P
CAS Number:
Molecular Weight:
861.89
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870287P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870287P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C1=NC(Cl)=NC(Cl)=N1)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

Packaging

5 mL Amber Glass Screw Cap Vial (870287P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heike Bakowsky et al.
Biochimica et biophysica acta, 1778(1), 242-249 (2007-10-30)
Carbohydrate moieties of the cellular glycocalyx have been suggested to play an important role in biological recognition processes during pathologic conditions, such as inflammation and cancer. Herein, we describe lectin-modified liposomes which might have potential for site-specific drug delivery during
G Bendas et al.
International journal of pharmaceutics, 181(1), 79-93 (1999-06-17)
In order to develop long-circulating immunoliposomes (IL), which combine sterical stabilization with a superior targetability, we have introduced a new methodology for attaching monoclonal antibodies directly onto the distal ends of liposome-grafted polyethylene glycol (PEG) chains. Therefore, we have synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service