Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870142C

Avanti

18:1 Dodecanylamine PE

Avanti Research - A Croda Brand 870142C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H101N2O9P
CAS Number:
Molecular Weight:
941.35
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870142C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870142C

concentration

10 mg/mL (870142C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN(C(CCCCCCCCCCC[NH3+])=O)[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C53H101N2O9P/c1-3-5-7-9-11-13-15-17-19-21-23-27-31-35-39-43-52(57)61-48-50(64-53(58)44-40-36-32-28-24-22-20-18-16-14-12-10-8-6-4-2)49-63-65(59,60)62-47-46-55-51(56)42-38-34-30-26-25-29-33-37-41-45-54/h17-20,50H,3-16,21-49,54H2,1-2H3,(H,55,56)(H,59,60)/b19-17-,20-18-/t50-/m1/s1

InChI key

HOOLPMPNZPBWNI-WSPRLCGESA-N

General description

18:1 Dodecanylamine PE, also referred as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine), is a head group modified functionalized lipid.

Packaging

5 mL Clear Glass Sealed Ampule (870142C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service