Skip to Content
MilliporeSigma
All Photos(1)

Documents

870111O

Avanti

Trioctanoin

>99% (TLC), Avanti Research - A Croda Brand

Synonym(s):

Trioctanoin, 5β-cholanic acid-3α,7α,12α-triol N-(2-sulphoethyl)-amide, sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

oil

packaging

package of 1 × 5 g

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (870111O-5G)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service