Skip to Content
MilliporeSigma
All Photos(1)

Documents

860573P

Avanti

C12 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/12:0)

Avanti Research - A Croda Brand 860573P, powder

Synonym(s):

3-O-sulfo-D-galactosyl-β1-1′-N-lauroyl-D-erythro-sphingosine (ammonium salt), (synthetic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H72N2O11S
CAS Number:
Molecular Weight:
741.03
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860573P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860573P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](OS([O-])(=O)=O)[C@H]([C@H]1CO)O.[NH4+]

InChI

1S/C36H69NO11S.H3N/c1-3-5-7-9-11-13-14-15-16-18-19-21-23-25-30(39)29(37-32(40)26-24-22-20-17-12-10-8-6-4-2)28-46-36-34(42)35(48-49(43,44)45)33(41)31(27-38)47-36;/h23,25,29-31,33-36,38-39,41-42H,3-22,24,26-28H2,1-2H3,(H,37,40)(H,43,44,45);1H3/b25-23+;/t29-,30+,31?,33-,34?,35?,36+;/m0./s1

InChI key

ZRHCDEVQWSMWDC-LNTPIPJUSA-N

Application

C12 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/12:0) is suitable for use as a standard in mass spectrometric analysis of sulfatides.

Biochem/physiol Actions

C12 Mono-Sulfo Galactosyl(β) Ceramide (d18:1/12:0) is a non-physical sulfatide. Accumulation of sulfatide impairs the activity of arylsulfatase A enzyme, which in turn leads to the development of genetic disease, metachromatic leukodystrophy.

Packaging

5 mL Amber Glass Screw Cap Vial (860573P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of plasma sulfatides by mass spectrometry: Utility for metachromatic leukodystrophy
Saville J, et al.
Analytica Chimica Acta, 955, 79-85 (2017)
Synthetic sulfogalactosylceramide (sulfatide) and its use for the mass spectrometric quantitative urinary determination in metachromatic leukodystrophies
Cui Y, et al.
Glycoconjugate Journal, 25(2), 147-155 (2008)
Mariachiara Bianco et al.
Food chemistry, 324, 126878-126878 (2020-04-29)
Ceramides (Cer) and cerebrosides are important sphingolipids (SL) involved in many biological processes. Herein, the SL content of yellow lupin seeds (Lupinus luteus) was determined by liquid-liquid extraction, mild alkaline hydrolysis (1 h at 37 °C) and reversed-phase liquid chromatography with negative
Michela Antonelli et al.
Analytical and bioanalytical chemistry, 412(2), 413-423 (2019-11-25)
The chemical composition of Cannabis sativa L. has been extensively investigated for several years; nevertheless, a detailed lipidome characterization is completely lacking in the literature. To achieve this goal, an extraction and enrichment procedure was developed for the characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service