Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860563P

Avanti

18:1-12:0 Biotin PC

Avanti Research - A Croda Brand 860563P, powder

Synonym(s):

1-oleoyl-2-[12-biotinyl(aminododecanoyl)]-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H89N4O10PS
CAS Number:
Molecular Weight:
945.28
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860563P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860563P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

Application

18:1-12:0 Biotin PC is suitable for use in the preparation of liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (860563P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Rab11-binding protein RELCH/KIAA1468 controls intracellular cholesterol distribution
Sobajima T, et al.
The Journal of Cell Biology, 217(5), 1777-1796 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service