Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860465P

Avanti

N-C24:1-desoxymethylsphinganine

Avanti Research - A Croda Brand 860465P, powder

Synonym(s):

N-nervonoyl-1-desoxymethylsphinganine (m17:0/24:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$528.00

$528.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$528.00

About This Item

Empirical Formula (Hill Notation):
C41H81NO2
CAS Number:
Molecular Weight:
620.09
UNSPSC Code:
12352211
NACRES:
NA.25

$528.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860465P-1mg)
pkg of 1 × 5 mg (860465P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860465P

lipid type

bioactive lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](CCCCCCCCCCCCCCC)(O)CNC(CCCCCCCCCCCCC/C=C/CCCCCCCC)=O

InChI

1S/C41H81NO2/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-41(44)42-39-40(43)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,40,43H,3-16,19-39H2,1-2H3,(H,42,44)/b18-17+/t40-/m1/s1

InChI key

LBLLNJQTYVMCAN-PESHEJLBSA-N

General description

N-C24:1-desoxymethylsphinganine Commonly referred to as 1-desoxymethyldihydroceramide (1-desoxyMeDHCer), is the N-acylated form of 1-desoxymethylsphinganine,[1] a potent inhibitor of sphingolipid metabolism. 1-deoxysphinganine (1-deoxySa) with N-acyl groups are referred as 1-deoxydihydroceramides and can have 16, 20 and 24 carbon chain.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860465P-1mg)
5 mL Amber Glass Screw Cap Vial (860465P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide synthase inhibition by fumonisin B1 causes accumulation of 1-deoxysphinganine a novel category of bioactive 1-deoxysphingoid bases and 1-deoxydihydroceramides biosynthesized by mammalian cell lines and animals
Zitomer NC, et al.
The Journal of Biological Chemistry, 284, 4786-4795 (2009)
Noemi Jiménez-Rojo et al.
Biophysical journal, 107(12), 2850-2859 (2014-12-18)
Ceramides and dihydroceramides are N-acyl derivatives of sphingosine and sphinganine, respectively, which are the major sphingoid-base backbones of mammals. Recent studies have found that mammals, like certain other organisms, also produce 1-deoxy-(dihydro)ceramides (1-deoxyDHCers) that contain sphingoid bases lacking the 1-hydroxyl-
Thematic Review Series: Sphingolipids. Biodiversity of sphingoid bases (?sphingosines?) and related amino alcohols
Pruett ST, et al.
Journal of Lipid Research, 49(8), 1621-1639 (2008)
Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service