Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860454P

Avanti

18:0 PA-d70

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-d70-sn-glycero-3-phosphate (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,750.00

$1,750.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,750.00

About This Item

Empirical Formula (Hill Notation):
C39H6O8PNaD70
CAS Number:
Molecular Weight:
797.41
UNSPSC Code:
12352100
NACRES:
NA.25

$1,750.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

1 ea of 10 mg (860454P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O)([O-])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=

General description

1,2-distearoyl-d70-sn-glycero-3-phosphate (sodium salt) (18:0 PA-d70) is a deuterated phosphatidic acid wherein 70 protons in distearoyl are replaced by deuterium isotope. Phosphatidic acid (PA) is a phospholipid located in the membrane.[1]

Biochem/physiol Actions

Phosphatidic acid (PA) plays an important role in signal transduction and membrane trafficking. It is also associated with cytoskeletal rearrangement and secretion. PA acts as an intermediate to produce storage and membrane lipids.[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Signaling functions of phosphatidic acid
Wang X, et al.
Progress in Lipid Research, 45(3), 250-278 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service