Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860320P

Avanti

16:0-18:1 PC-d82

Avanti Research - A Croda Brand 860320P, powder

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine-d82

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$871.38

$871.38


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$871.38

About This Item

Empirical Formula (Hill Notation):
C42D82NO8P
CAS Number:
Molecular Weight:
842.58
UNSPSC Code:
51191904
NACRES:
NA.25

$871.38


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860320P-1mg)
pkg of 1 × 5 mg (860320P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860320P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OC([2H])(C([N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])[2H])[2H])(OC([C@]([2H])(OC(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C(/C([2H])=C([2H])\C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H

General description

16:0-18:1 PC-d82 is a deuterated phosphocholine.

Application

16:0-18:1 PC-d82 has been used for the preparation of giant unilamellar vesicles (GUVs).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860320P-1mg)
5 mL Amber Glass Screw Cap Vial (860320P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquidity Is a Critical Determinant for Selective Autophagy of Protein Condensates
Yamasaki A, et al.
Molecular Cell (2020)
Akinori Yamasaki et al.
Molecular cell, 77(6), 1163-1175 (2020-01-30)
Clearance of biomolecular condensates by selective autophagy is thought to play a crucial role in cellular homeostasis. However, the mechanism underlying selective autophagy of condensates and whether liquidity determines a condensate's susceptibility to degradation by autophagy remain unknown. Here, we
Debasis Das et al.
Nature communications, 11(1), 231-231 (2020-01-15)
The opening of a fusion pore during exocytosis creates the first aqueous connection between the lumen of a vesicle and the extracellular space. Soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs) mediate the formation of these dynamic structures, and their kinetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service