Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860131P

Avanti

GM2 Sphingosine (d18:1)

new

Avanti Research - A Croda Brand

Synonym(s):

GM2 Ganglioside (GalNAcb1,4(Neu5Aca2,3)Galb1,4Glcb-Sphingosine(d18:1))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H87N3O25
Molecular Weight:
1118.22
UNSPSC Code:
12352211
NACRES:
NA.79

biological source

synthetic

Assay

>99 (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](O)[C@H]([C@H](O[C@@H]2[C@H](O[C@@]3(C[C@H](O)[C@H]([C@H]([C@H](O)[C@H](O)CO)O3)NC(C)=O)C(O)=O)[C@@H](O)[C@H](O[C@H]4C(O)[C@@H](O)[C@H](OC[C@@H]([C@H](O)/C=C/CCCCCCCCCCCCC)N)O[C@@H]4CO)O[C@@H]2CO)O[C@@H]1CO)NC(C)=O

Application

Lipids research

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service