Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

857136C

Avanti

18:1 BMP (R,R)

sn-(1-oleoyl-2-hydroxy)-glycerol-3-phospho-sn-3′-(1′-oleoyl-2′-hydroxy)-glycerol (ammonium salt), chloroform

Synonym(s):

bis(monoacyl-glycerol)phosphate, lysobisphosphatidic acid; LBPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO10P
CAS Number:
Molecular Weight:
792.07
UNSPSC Code:
12352211

Assay

>99% (BMP (contains 30% positional isomers), TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL (857136C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857136C

concentration

5 mg/mL (857136C-5mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OC[C@]([H])(O)COC(CCCCCCC/C=C\CCCCCCCC)=O)([O-])=O)(O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

General description

18:1 BMP (R,R)/Lysobisphosphatidic acid (LBPA) is an unconventional phospholipid. It is usually present in late endosomes. It is also termed as bis(monoacyl-glycerol)phosphate.

Biochem/physiol Actions

18:1 BMP (R,R)/Lysobisphosphatidic acid (LBPA) can modulate endosomal cholesterol levels. It participates in cholesterol transport. LBPA helps to regulate the cholesterol storage capacity of endosomes.

Packaging

5 mL Clear Glass Sealed Ampule (857136C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The stereochemical configuration of lysobisphosphatidic acid from rat liver, rabbit lung and pig lung
Joutti A, et al.
Biochimica et Biophysica Acta, Lipids and Lipid Metabolism, 450(2), 206-209 (1976)
Lysobisphosphatidic acid controls endosomal cholesterol levels
Chevallier J, et al.
The Journal of Biological Chemistry, 283(41), 27871-27880 (2008)
Elena Zaitseva et al.
PLoS pathogens, 6(10), e1001131-e1001131 (2010-10-16)
Many enveloped viruses invade cells via endocytosis and use different environmental factors as triggers for virus-endosome fusion that delivers viral genome into cytosol. Intriguingly, dengue virus (DEN), the most prevalent mosquito-borne virus that infects up to 100 million people each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service