Skip to Content
MilliporeSigma
All Photos(2)

Documents

855774P

Avanti

2-18:0 Lyso PC

Avanti Research - A Croda Brand 855774P, powder

Synonym(s):

2-stearoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H54 NO7P
CAS Number:
Molecular Weight:
523.68
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (855774P-1mg)
pkg of 1 × 10 mg (855774P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855774P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)CO)=O

Packaging

5 mL Amber Glass Screw Cap Vial (855774P-10mg)
5 mL Amber Glass Screw Cap Vial (855774P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service