Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850706C

Avanti

16:1 PE

1,2-dipalmitoleoyl-sn-glycero-3-phosphoethanolamine, chloroform

Synonym(s):

1,2-di-(9Z-hexadecenoyl)-sn-glycero-3-phosphoethanolamine; PE(16:1(9Z)/16:1(9Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$325.48

$325.48


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$325.48

About This Item

Empirical Formula (Hill Notation):
C37H70NO8P
CAS Number:
Molecular Weight:
687.93
UNSPSC Code:
51191904
NACRES:
NA.25

$325.48


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850706C-25mg)
pkg of 2 × 4 mL (850706C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850706C

concentration

10 mg/mL (850706C-25mg)
25 mg/mL (850706C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCC)=O)COC(CCCCCCC/C=C\CCCCCC)=O

InChI

1S/C37H70NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-36(39)43-33-35(34-45-47(41,42)44-32-31-38)46-37(40)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h13-16,35H,3-12,17-34,38H2,1-2H3,(H,41,42)/b15-13-,16-14-/t35-/m1/s1

InChI key

PGPMCWZMPPZJML-NAFNZUQFSA-N

General description

Phosphatidylethanolamine (PE), a phospholipid is found abundantly in mammalian cells.[1]

Application

1,2-dipalmitoleoyl-sn-glycero-3-phosphoethanolamine (16:1 PE) may be used in the preparation of vesicles[2] and in micelle preparation for cell experiments.[3]

Biochem/physiol Actions

Phosphatidylethanolamine (PE) serves as a lipid chaperone that supports in the folding of membrane proteins.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850706C-200mg)
5 mL Clear Glass Sealed Ampule (850706C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atanas V Koulov et al.
Biochimica et biophysica acta, 1564(2), 459-465 (2002-08-15)
Cationic, triple-chain amphiphiles promote vesicle fusion more than structurally related double-chain or single-chain analogues. Two types of vesicle fusion experiments were conducted, mixing of oppositely charged vesicles and acid-triggered self-fusion of vesicles composed of cationic amphiphile and anionic cholesteryl hemisuccinate
Tomoyuki Nishizaki
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 46(5), 1985-1998 (2018-05-04)
Phosphatidylethanolamine, a component of the plasma membrane, regulates diverse cellular processes. The present study investigated the role of 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE) in the trafficking of the glucose transporter GLUT4 and the glucose homeostasis. Monitoring of GLUT4 trafficking, GLUT4 internalization assay, and
Dhaval Patel et al.
Oxidative medicine and cellular longevity, 2017, 4829180-4829180 (2017-08-09)
Phosphatidylethanolamine (PE) is the second most abundant phospholipid in mammalian cells. PE comprises about 15-25% of the total lipid in mammalian cells; it is enriched in the inner leaflet of membranes, and it is especially abundant in the inner mitochondrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service