Skip to Content
MilliporeSigma
All Photos(2)

Documents

850527P

Avanti

Mal(11.1)

dodecan-2-yloxy-β-D-maltoside, powder

Synonym(s):

2-dodecyl-Β-D-maltopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46O11
CAS Number:
Molecular Weight:
510.62
UNSPSC Code:
12161902
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850527P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850527P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OC(C)CCCCCCCCCC)[C@H](O)[C@@H](O)[C@@H]1O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2

General description

This new class of detergents contain a short, branched alkyl chain at the interface between the polar head and the apolar tail. This design mimics the second aliphatic chain of lipid molecules and reduces water penetration, thereby increasing the hydrophobicity within the interior of the micelle. A detergent architecture of this style allows formation of smaller, more compact micelles which provides improved performance in the stabilization and crystallization of integral membrane proteins.

Packaging

20 mL Clear Glass Screw Cap Vial (850527P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Xu Hong et al.
Langmuir : the ACS journal of surfaces and colloids, 26(11), 8690-8696 (2010-03-18)
A challenging requirement for structural studies of integral membrane proteins (IMPs) is the use of amphiphiles that replicate the hydrophobic environment of membranes. Progress has been impeded by the limited number of useful detergents and the need for a deeper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service