Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850501P

Avanti

CHAPSO

Avanti Research - A Croda Brand 850501P, powder

Synonym(s):

3-[(3-Cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$213.21

$213.21


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$213.21

About This Item

Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
UNSPSC Code:
12161902
NACRES:
NA.25

$213.21


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>98% (TLC)

form

powder

mol wt

630.88 g/mol

packaging

pkg of 1 × 1 g (850501P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850501P

technique(s)

protein quantification: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12C(CC[C@@H]2[C@@H](CCC(NCCC[N+](C)(CC(O)CS(=O)([O-])=O)C)=O)C)C3[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)C3C[C@@H]1O

InChI

1S/C32H58N2O8S/c1-20(7-10-29(39)33-13-6-14-34(4,5)18-23(36)19-43(40,41)42)24-8-9-25-30-26(17-28(38)32(24,25)3)31(2)12-11-22(35)15-21(31)16-27(30)37/h20-28,30,35-38H,6-19H2,1-5H3,(H-,33,39,40,41,42)/t20-,21+,22-,23?,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

GUQQBLRVXOUDTN-XOHPMCGNSA-N

General description

3-(cholamidopropyl)(dimethylammonio)-2-hydroxy-1-propanesulfonate (CHAPSO) is a detergent and a bile salt analog.[1]
Unique detergent with high CMC that solubilizes membrane-bound proteins in their native state. Capable of solubilizing opiate receptor to a state exhibiting reversible binding of opiates. Has higher solubility than CHAPS because of its more polar head group. Micellar average MW: 7000 Aggregation number: 11

Application

3-(cholamidopropyl)(dimethylammonio)-2-hydroxy-1-propanesulfonate (CHAPSO) may be used in bicelle preparation for β2-adrenergic receptor reconstitution.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (850501P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantin E Komolov et al.
Cell, 169(3), 407-421 (2017-04-22)
The phosphorylation of agonist-occupied G-protein-coupled receptors (GPCRs) by GPCR kinases (GRKs) functions to turn off G-protein signaling and turn on arrestin-mediated signaling. While a structural understanding of GPCR/G-protein and GPCR/arrestin complexes has emerged in recent years, the molecular architecture of
W F Simonds et al.
Proceedings of the National Academy of Sciences of the United States of America, 77(8), 4623-4627 (1980-08-01)
Receptors that reversibly bind opiates and opioid peptides have been solubilized from brain and neuroblastoma-glioma hybrid cell NG108-15 membranes. Active receptors are specifically solubilized with a new type of detergent 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonate, which is a zwitterionic derivative of cholic acid. The
Yasser Qutub et al.
The journal of physical chemistry. B, 114(13), 4529-4535 (2010-03-20)
To address the interactions of hemin with phospholipid bilayers, we introduce hemin to a solution of dimyristoylphosphatidylcholine (DMPC), a long chain phospholipid, and 3-(cholamidopropyl)(dimethylammonio)-2-hydroxy-1-propanesulfonate (CHAPSO), a detergent, in which vesicles form at around 37 degrees C. We show that vesicles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service