Skip to Content
MilliporeSigma
All Photos(2)

Documents

850481P

Avanti

16:0-18:0 (9-10BR) PC

1-palmitoyl-2-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-hexadecanoyl-2-(9,10-dibromooctadecanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8PBr2
CAS Number:
Molecular Weight:
919.88
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (850481P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850481P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCC(Br)C(Br)CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

General description

16:0-18:0 (9-10BR) PC (1-palmitoyl-2-(9,10-dibromo) stearoyl-sn-glycero-3-phosphocholine) is a brominated phosphatidylcholine lipid.

Application

16:0-18:0 (9-10BR) PC (1-palmitoyl-2-(9,10-dibromo) stearoyl-sn-glycero-3-phosphocholine) has been used in large unilamellar vesicle preparation. It may be used as a component in large unilamellar liposomes for parallax analysis and in lipid vesicle preparation for tryptophan (Trp) penetration depth experiments/bromine quenching experiments.

Biochem/physiol Actions

16:0-18:0 (9-10BR) PC (1-palmitoyl-2-(9,10-dibromo)stearoyl-sn-glycero-3-phosphocholine) serves as a deep quencher.

Packaging

5 mL Clear Glass Sealed Ampule (850481P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thai Leong Yap et al.
The Journal of biological chemistry, 290(2), 744-754 (2014-11-28)
Mutations in glucocerebrosidase (GCase), the enzyme deficient in Gaucher disease, are a common genetic risk factor for the development of Parkinson disease and related disorders, implicating the role of this lysosomal hydrolase in the disease etiology. A specific physical interaction
Henriette Aksnes et al.
The Journal of biological chemistry, 292(16), 6821-6837 (2017-02-16)
Nα-Acetyltransferase 60 (Naa60 or NatF) was recently identified as an unconventional N-terminal acetyltransferase (NAT) because it localizes to organelles, in particular the Golgi apparatus, and has a preference for acetylating N termini of the transmembrane proteins. This knowledge challenged the
John M Harrington et al.
PloS one, 7(9), e44384-e44384 (2012-09-13)
The bloodstream developmental forms of pathogenic African trypanosomes are uniquely susceptible to killing by small hydrophobic peptides. Trypanocidal activity is conferred by peptide hydrophobicity and charge distribution and results from increased rigidity of the plasma membrane. Structural analysis of lipid-associated
Candace M Pfefferkorn et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2821-2828 (2015-08-25)
Apolipoproteins are essential human proteins for lipid metabolism. Together with phospholipids, they constitute lipoproteins, nm to μm sized particles responsible for transporting cholesterol and triglycerides throughout the body. To investigate specific protein-lipid interactions, we produced and characterized three single-Trp containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service