Skip to Content
MilliporeSigma
All Photos(2)

Documents

850479C

Avanti

16:0-18:0(4,5-dibromo) PC

Avanti Research - A Croda Brand 850479C

Synonym(s):

1-palmitoyl-2-stearoyl(4,5)dibromo-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82Br2NO8P
CAS Number:
Molecular Weight:
919.88
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (850479C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850479C

concentration

10 mg/mL (850479C-10mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC(Br)C(Br)CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

General description

16:0-18:0(4,5-dibromo) PC
(1-palmitoyl-2-stearoyl (4,5) dibromo-sn-glycero-3-phosphocholine) is a lipid made up of phosphocholine backbone, palmitoyl and stearoyl fatty acid chains. The 4th and 5th carbons of stearoyl group has bromo substitution.

Application

16:0-18:0(4,5-dibromo) PC (1-palmitoyl-2-stearoyl (4,5) dibromo-sn-glycero-3-phosphocholine) may be used as a component in the vesicles for quenching experiments.

Biochem/physiol Actions

16:0-18:0(4,5-dibromo) PC (1-palmitoyl-2-stearoyl (4,5) dibromo-sn-glycero-3-phosphocholine) can be used to prepare vesicles.

Packaging

5 mL Clear Glass Sealed Ampule (850479C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cari M Anderson et al.
The journal of physical chemistry. B, 123(1), 170-179 (2018-11-28)
The interactions between small molecules and lipid bilayers play a critical role in the function of cellular membranes. Understanding how a small molecule interacts with the lipid bilayer differently based on its charge reveals primordial mechanisms of transport across membranes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service