Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

840432P

Avanti

06:0 PG

1,2-dihexanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

1,2-dicaproyl-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(6:0/6:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O10PNa
CAS Number:
Molecular Weight:
464.42
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840432P-10mg)
pkg of 1 × 200 mg (840432P-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840432P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCC)COC(=O)CCCCC)OCC(O)CO

InChI

1S/C18H35O10P.Na/c1-3-5-7-9-17(21)25-13-16(28-18(22)10-8-6-4-2)14-27-29(23,24)26-12-15(20)11-19;/h15-16,19-20H,3-14H2,1-2H3,(H,23,24);/q;+1/p-1/t15?,16-;/m1./s1

InChI key

URXZWMWYOKBMMW-NPSJIHRQSA-M

General description

06:0 PG (1,2-dihexanoyl-sn-glycero-3-phospho-(1′-rac-glycerol), a negatively charged phospholipid is hygroscopic and not sensitive to light.

Application

06:0 PG (1,2-dihexanoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) may be used as a standard and a short/odd-fatty acyl chain analog to spike the lipid mixture for the quantification of a natural lipid mixture (egg PG). It may also be used in micelles for lipid-binding studies.

Biochem/physiol Actions

Phosphatidylglycerol (PG) participates in photosynthetic electron transport.

Packaging

20 mL Clear Glass Screw Cap Vial (840432P-200mg)
5 mL Clear Glass Sealed Ampule (840432P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja A Dames et al.
The Journal of biological chemistry, 286(42), 36907-36920 (2011-08-19)
Dictyostelium Formin C (ForC) is involved in the regulation of local actin cytoskeleton reorganization (e.g. during cellular adhesion or migration). ForC contains formin homology 2 and 3 (FH2 and -3) domains and an N-terminal putative GTPase-binding domain (GBD) but lacks
Koichi Kobayashi et al.
Plant cell reports, 34(4), 631-642 (2014-12-06)
With phosphate deficiency, the role of phosphatidylglycerol is compensated by increased glycolipid content in thylakoid membrane biogenesis but not photosynthetic electron transport in Arabidopsis chloroplasts. In plants and cyanobacteria, anionic phosphatidylglycerol (PG) is the only major phospholipid in thylakoid membranes
Tommy Hofmann et al.
Chemistry and physics of lipids, 223, 104782-104782 (2019-06-10)
In recent years, lipid quantification gained importance. In most cases, this is achieved by spiking the lipid mixture with deuterated standard lipids or lipid analogues that differ in chain length when compared with the natural lipid components. Usually, conventional ESI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service