Skip to Content
MilliporeSigma
All Photos(2)

Documents

810890P

Avanti

Fluorescent DOTAP

1-oleoyl-2-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-3-trimethylammonium propane (chloride salt), powder

Synonym(s):

18:1-06:0 NBD DOTAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60ClN5O7
CAS Number:
Molecular Weight:
710.34
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 50 μg (810890P-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810890P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(C[N+](C)(C)C)(OC(CCCCCNC1=CC=C([N+]([O-])=O)C2=NON=C12)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Cl-]

General description

N-(1-(2,3-dioleoyloxy)propyl)-N,N,N-trimethylammonium (DOTAP) is a cationic lipid, which forms strong complexes with DNA. It aids the cellular delivery of RNA and DNA. DOTAP acts as a transfection agent.

Application

Fluorescent DOTAP has been used for the preparation of liposomes loaded with gold and verteporfin (VP).

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810890P-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurons: Methods and Applications for the Cell Biologist (2003)
Controlled gene and drug release from a liposomal delivery platform triggered by X-ray radiation
Deng W, et al.
Nature Communications, 9(1), 2713-2713 (2018)
Phospholipid-cationic lipid interactions: influences on membrane and vesicle properties
Campbell RB, et al.
Biochimica et Biophysica Acta - Biomembranes, 1512(1), 27-39 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service