Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810351P

Avanti

22-NBD 25-hydroxycholesterol

Synonym(s):

22-[N-(7-nitro-2-1,3-benzoxadiazol-4-yl)]-25-hydroxy-23-azacholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H46N4O5
CAS Number:
Molecular Weight:
566.73
UNSPSC Code:
12352211
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810351P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

fluorescent lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@H](CN(CC(C)(C)O)c1ccc([N+]([O-])=O)c2nonc12)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Application

Fluorescence

Packaging

5 mL Amber Glass Screw Cap Vial (810351P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service