Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

810225P

Avanti

18:1 Dansyl PS

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phospho-L-serine-N-(5-dimethylamino-1-naphthalenesulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$710.70

$710.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$710.70

About This Item

Empirical Formula (Hill Notation):
C54H95N4O12PS
CAS Number:
Molecular Weight:
1055.39
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$710.70


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810225P-1mg)
pkg of 5 × 1 mg (810225P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylserine (PS) belongs to the class of glycerophospholipids and is generally found in the inner leaflet of the plasma membrane and in endocytic membranes.[1] Dansyl ((dimethyl amino)naphthalene-1-sulfonYL)) group produces blue or blue-green fluorescent adduct and is used in labeling techniques for studying protein folding and dynamics.(5)

Application

18:1 Dansyl PS may be used for preparation of liposomes for sedimentation assays, visualising and other fluorometric analysis like FRET.[2]

Biochem/physiol Actions

Phosphatidylserine (PS) plays a vital role in cell cycle signaling and engulfing apoptotic cells.[3] It is also known to activate the blood coagulation process acting as a pro-coagulant surface.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (810225P-1mg)
5 mL Amber Glass Screw Cap Vial (810225P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supplemental Data ESCRT-I Core and ESCRT-II GLUE Domain Structures Reveal Role for GLUE in Linking to ESCRT-I and Membranes
Advances in Planar Lipid Bilayers and Liposomes, 125(1), 99-111 (2011)
Laurent Meertens et al.
Cell host & microbe, 12(4), 544-557 (2012-10-23)
Dengue viruses (DVs) are responsible for the most medically relevant arboviral diseases. However, the molecular interactions mediating DV entry are poorly understood. We determined that TIM and TAM proteins, two receptor families that mediate the phosphatidylserine (PtdSer)-dependent phagocytic removal of
Barry R Lentz
Progress in lipid research, 42(5), 423-438 (2003-06-20)
This article addresses the role of platelet membrane phosphatidylserine (PS) in regulating the production of thrombin, the central regulatory molecule of blood coagulation. PS is normally located on the cytoplasmic face of the resting platelet membrane but appears on the
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service