Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810223P

Avanti

C12-NBD Glucosyl Ceramide

Avanti Research - A Croda Brand

Synonym(s):

N-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-D-glucosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$1,915.80
810223P-1MG
$2,380.00

$1,915.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
$1,915.80
810223P-1MG
$2,380.00

About This Item

Empirical Formula (Hill Notation):
C42H71N5O11
CAS Number:
Molecular Weight:
822.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,915.80


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810223P-1MG)
pkg of 1 × 250 μg (810223P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Application

C12-NBD Glucosyl Ceramide is suitable for liposome preparation.[1]

Biochem/physiol Actions

Glucosyl ceramide (GlcCer) is produced by GlcCer synthase from ceramide in the cytosolic membranes of Golgi. In the luminal Golgi membrane, GlcCer participates in the generation of glycosphingolipids.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810223P-1MG)
5 mL Amber Glass Screw Cap Vial (810223P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of chemically synthesized saposin C on glucosylceramide-beta-glucosidase
Yoneshige A, et al.
Clinical Biochemistry, 48(16-17), 1177-1180 (2015)
Glycosphingolipid synthesis requires FAPP2 transfer of glucosylceramide
D?Angelo G, et al.
Nature, 449(7158), 62-62 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service